Learn More
R&D Systems™ Human IL-23R (AAM44229) VersaClone cDNA
IL-23R; interleukin 23 receptor; interleukin-23 receptor
Brand:  R&D Systems™ RDC1255
Encompass
Product Code. 15741245
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Human IL-23 R (AAM44229) VersaClone cDNA clone contains the complete ORF for IL-23R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-23R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
hIL23R
10 μg at 0.2 μg/μL
1930bp
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Human, Mouse
AAM44229
cDNA is provided in 10mM Tris-Cl, pH 8.5
149233, 209590
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans.