missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-(+)-3-Chloro-1-phenyl-1-propanol, 98%

Product Code. 15441868
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 100306-33-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.636
MDL Number MFCD00075128
InChI Key JZFUHAGLMZWKTF-SECBINFHSA-N
Synonym r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738
PubChem CID 642409
IUPAC Name (1R)-3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O

Specifications

Melting Point 58°C to 60°C
Quantity 1 g
Beilstein 5250766
Optical Rotation +26° (c=1 in Chloroform)
Formula Weight 170.64
Percent Purity 98%
Chemical Name or Material (R)-(+)-3-Chloro-1-phenyl-1-propanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.