missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-4-hydroxybutyramide, 97%, Thermo Scientific™

Product Code. 15439337
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256482-94-6
Molecular Formula C9H18N2O4
Molecular Weight (g/mol) 218.253
MDL Number MFCD00270250
InChI Key BYCKHNZSBNGBQL-ZCFIWIBFSA-N
Synonym n-boc-d-asparaginol, boc-d-asparaginol, r-3-boc-amino-4-hydroxybutyramide, n-boc-r-3-amino-4-hydroxybutanoic acid amide, tert-butyl n-2r-1-carbamoyl-3-hydroxypropan-2-yl carbamate
PubChem CID 51387393
IUPAC Name tert-butyl N-[(2R)-4-amino-1-hydroxy-4-oxobutan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)CO

Specifications

Formula Weight 218.25
Percent Purity 97%
Quantity 1 g
Chemical Name or Material (R)-3-(Boc-amino)-4-hydroxybutyramide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'