missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-(Anilinomethyl)pyrrolidine, 98%, Thermo Scientific™

Product Code. 11389575
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the determination of the absolute configuration of amines and α-amino acids by 1h nmr of (r)-o-aryllactic acid amides.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from air, carbon dioxide and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68295-45-4
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD01862117
InChI Key MCHWKJRTMPIHRA-LLVKDONJSA-N
Synonym r-n-pyrrolidin-2-ylmethyl aniline, r---2-anilinomethylpyrrolidine, r---2-anilinomethyl pyrrolidine, n-2r-pyrrolidin-2-yl methyl aniline, n-2r-pyrrolidin-2-ylmethyl aniline, r-2-anilinomethyl pyrrolidine, r-2-phenylaminomethyl-pyrrolidine, r-2-phenylamino-methyl pyrrolidine
PubChem CID 6950383
IUPAC Name N-[[(2R)-pyrrolidin-2-yl]methyl]aniline
SMILES C1CC(NC1)CNC2=CC=CC=C2

Specifications

Density 1.046
Boiling Point 120°C to 122°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.575
Quantity 1 g
UN Number UN2735
Beilstein 4839749
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Optical Rotation −20° (c=1 in Ethanol)
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-(Anilinomethyl)pyrrolidine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.