missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminooctane, 98+%, ee 98+%, Thermo Scientific Chemicals

Product Code. 11334216
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34566-05-7
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD01091017
InChI Key HBXNJMZWGSCKPW-MRVPVSSYSA-N
Synonym r-2-aminooctane, r-2-octylamine, 2r-2-octanamine, 2-octanamine, 2r, r---2-aminooctane, unii-rjm666k1lk, 2r-octan-2-amine, rjm666k1lk, l-2-aminooctane, --2-octylamine
PubChem CID 2734519
IUPAC Name (2R)-octan-2-amine
SMILES CCCCCCC(C)N

Specifications

Melting Point <-20°C
Density 0.77
Boiling Point 163°C to 165°C
Flash Point 50°C (122°F)
Refractive Index 1.4235
Quantity 1 g
UN Number UN2734
Beilstein 1719320
Sensitivity Air Sensitive
Formula Weight 129.25
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (R)-(-)-2-Aminooctane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'