Learn More
SVL™ Socket/Screw Joint Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
Brand:  SVL 670-00
 View more versions of this product
Product Code. 15547740
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Features an SVL™ screwthread and Quickfit™ ground glass socket
  • Thread accepts SVL™ screw caps and couplings. Socket fits Quickfit™ cones
  • Manufactured from temperature and chemical resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SVL™ and Quickfit™ Glassware
Borosilicate Glass
Socket/screw thread ground joint adapter
15