Learn More
Quickfit™ Pear-Shaped Glass Funnel with Glass Key and Stem with Cone
For use in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
Brand:  Quickfit D2/12
 View more versions of this product
Product Code. 12438846
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For use in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
  • Complies with ISO 4800 Type 2
  • Funnel stem with cone to allow easy connection to Quickfit™ flasks etc
  • Stopcock features interchangeable glass key
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50mm
Borosilicate Glass
19/26
1PK
Separating funnel
50 mL
ISO 4800
19/26
1.5mm; Glass