Learn More
Quickfit™ Glass Conical Funnel with Semi-Micro Glass Sintered Disc
Brand: Scilabware SF4T30
This item is not returnable.
View return policy
Description
- Ideal for filtering semi-micro quantities of material
- Fitted with borosilicate glass sintered disc
- Suitable for use with Quickfit™ MD range of receivers
Specifications
Semi-micro filter funnel | |
10/19 | |
10mm | |
3 |
Borosilicate Glass | |
Quickfit™ MD: range of receivers | |
Sintered glass disc |