Learn More
Quickfit™ Flat Flange Joint
Ideal for use as pipeline joints or in the construction of flat flange lids for use with wide neck reaction flasks.
Brand:  Quickfit FG75
 View more versions of this product
Product Code. 15587840
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Highest quality flat flange joints
  • Precision ground for leak free performance
  • Choice of sizes up to 100mm bore
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.17in.
4.4mm