Learn More
Hellma™ Quartz Suprasil™ Absorption Cells
Thanks to their stability, maximum precision and reliability when used for absorbance measurements, Hellma cuvettes work exceptionally well in a wide range of areas in the lab.
Brand: Hellma™ 119-10-40
This item is not returnable.
View return policy
Specifications
Absorption Cell | |
Quartz Suprasil | |
1Each | |
200 to 2500 nm | |
12.5 mm |
3.5 mL | |
9.5mm | |
10 mm | |
49.5 mm | |
For Magnetic Stirrer |