missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele™ Quarter Segment Test Tube Insert
311.12 € - 356.03 €
Specifications
Product Type | Quarter Segment Test Tube Insert |
---|---|
Depth (Metric) | 320 mm |
Width (Metric) | 200 mm |
Quantity | 1 Each |