Learn More
Macherey Nagel Qualitative Analysis Test Papers, MN 615
Widely used for routine separations.
6.30 € - 7.70 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10758460
|
Macherey Nagel
431001 |
10 mm |
6.30 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10391255
|
Macherey Nagel
4310015 |
15 mm |
6.30 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10187374
|
Macherey Nagel
431002 |
20 mm |
6.30 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10499634
|
Macherey Nagel
4310025 |
25 mm |
6.30 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10320415
|
Macherey Nagel
431003 |
30 mm |
6.30 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10402275
|
Macherey Nagel
4310082 |
82 mm |
7.70 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10176087
|
Macherey Nagel
4310085 |
85 mm |
7.70 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Qualitative filter papers widely used for routine separations
- Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
- Most widely used general purpose filter paper with medium retention and flow rate
- This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids
Specifications
Disc | |
MN 615 | |
100/Pk. |