Learn More
Macherey Nagel Qualitative Analysis Test Papers
For the qualitative determination of ions and chemical compounds.
Brand:  Macherey Nagel 90732
Product Code. 10358461
  • 46.15 € / Pack of 200
Explore more special offers
Description
Description
  • Test papers allow the qualitative determination of ions and chemical compounds
  • They are used to detect whether compounds tested for are present above the specific detection limit
  • Some of the papers have specific applications
Specifications
Specifications
Silver ions (AG +)
200 x 20 mm x 70 mm strips
Qualitative analysis test strips