Learn More
MP Biomedicals™ qPCR & Go SYBR Hi-ROX Kit
Fast and sensitive qPCR SYBR Master Mix for most ABI platforms.
Brand: MP Biomedicals™ 11EBI01050
This item is not returnable.
View return policy
Description
- The qPCR & Go SYBR™ Hi-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
- A combination of the latest advances in buffer chemistry and enhancers, together with an antibody-mediated hot-start DNA polymerase system, ensures that the qPCR & Go SYBR™ Hi-ROX Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
- For ease-of-use and added convenience, qPCR & Go SYBR™ Hi-ROX is provided as a 2x mastermix containing all the components necessary for real-time PCR, including the SYBR™Green I dye, dNTPs, stabilisers and enhancers.
- The kit consists of a ready-to-use premix, only primers and template need to be added.
Specifications
-20°C | |
qPCR Kit | |
Real Time PCR (qPCR) |
Kit | |
5 x 1 mL | |
500 Reactions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.