missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine-1-carboximidamide hydroiodide, 97%, Thermo Scientific™

Product Code. 10653593
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 102392-83-6
Molecular Formula C5H12IN3
Molecular Weight (g/mol) 241.076
MDL Number MFCD00114285
InChI Key XPASGQSDLPDWNV-UHFFFAOYSA-N
Synonym pyrrolidine-1-carboximidamide hydroiodide, pyrrolidinecarboxamidine, iodide, c5h11n3.hi
PubChem CID 2794596
IUPAC Name pyrrolidine-1-carboximidamide;hydroiodide
SMILES C1CCN(C1)C(=N)N.I

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 241.07
Percent Purity 97%
Chemical Name or Material Pyrrolidine-1-carboximidamide hydroiodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'