missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, puriss. p.a., ACS reagent, Honeywell
259.76 € - 313.66 €
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.102 |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15634010
|
Honeywell Riedel-de Haën
33553-500ML |
500 mL |
313.66 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624010
|
Honeywell Riedel-de Haën
33553-1L |
1 L |
259.76 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
110-86-1 | |
0.124 | |
899°F | |
MFCD00011732 | |
UN1282 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
≥99.5% (GC) | |
Pyridine |
0.978g/mL at 25°C | |
17°C (62.6°F) | |
C5H5N | |
20 mmHg (25°C), 10 mmHg (13.2°C) | |
103233 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1g/mol | |
ACS/puriss. p.a. |
Safety and Handling
P210-P280-P305 + P351 + P338
Danger
missing translation for 'einecsNumber' : 203-809-9
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title