missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Certified AR for Analysis, Fisher Chemical™

Product Code. 10419570
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 115 deg.C, CHEBI:16227, Colorless, 79.1g/mol, JUJWROOIHBZHMG-UHFFFAOYSA-N, pyridine, -42 deg.C, 11732, 79.102

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number 11732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

pH 8.5
Melting Point -42°C
Color Colorless
Boiling Point 115°C
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 20mbar at 20°C
Viscosity 0.95 mPaS at 20°C
Formula Weight 79.1g/mol
compliance-icons compliance-icons
Product Identifier
  • Pyridine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'