missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, ≥99%, Reagent Grade, Honeywell™
Reagent Grade,≥99%
55.02 € - 366.96 €
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.102 |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15658040
|
Honeywell Research Chemicals
P57506-250ML |
250 mL | Glass Bottle |
55.02 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15668040
|
Honeywell Research Chemicals
P57506-500ML |
500 mL | Glass Bottle |
96.52 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15638040
|
Honeywell Research Chemicals
P57506-1L |
1 L | Glass Bottle |
166.79 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15648040
|
Honeywell Research Chemicals
P57506-2.5L |
2.5 L | Glass Bottle |
366.96 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
8.8 (at 20°C) | |
110-86-1 | |
0.978g/mL at 25°C | |
0.124 | |
899°F | |
Liquid | |
MFCD00011732 | |
UN1282 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1g/mol | |
Pyridine |
-42°C | |
Colorless | |
115°C (1013 hPa) | |
17°C (62.6°F) | |
Unpleasant | |
C5H5N | |
10mmHg (13.2°C), 20mmHg (25°C) | |
103233 | |
Fully miscible in water | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
≥99% |
Safety and Handling
P210-P280-P305 + P351 + P338
missing translation for 'dotInformation' : class 3 - PG 2 - Pyridine
missing translation for 'rtecsNumber' : 203-809-9
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Pyridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : pyridine