missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ PBT Screwcaps
Use with reagent bottles, culture tubes and other screwtop bottles. Pyrex™ PBT Screwcaps come in a wide range of sizes for your laboratory needs. Sealing discs provide inert inner seal, resistance to moisture vapor, and are great for culture work.
Brand: Pyrex 4503/30
103.55 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- PTFE faced sealing disc for chemical resistance
- Withstands hot air sterilization to 180deg.C
- Autoclavable
Specifications
Screw cap | |
PBT | |
30 mm | |
Autoclavable | |
Screw Cap |
PTFE faced rubber insert | |
20/PK | |
PTFE faced Rubber | |
Culture tubes |