missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Culture Flask

Ideal for use in preparation of cultures

Brand:  Pyrex 1135/14D

255.55 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12514917

  • 276.00 € / Pack of 10
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compatible with shaker equipment
  • Long, rimless tube neck for use with closures
  • Manufactured from Pyrex™ borosilicate glass with white enamel marking
TRUSTED_SUSTAINABILITY
Specifications

Specifications

38 mm
Borosilicate Glass
10
Conical
85 mm
250 mL
148 mm
Culture Flask
Flat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers