missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Culture Flask

Ideal for use in preparation of cultures

Brand:  Pyrex 1135/20D

124.45 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12504917

  • 135.00 € / Pack of 5
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compatible with shaker equipment
  • Long, rimless tube neck for use with closures
  • Manufactured from Pyrex™ borosilicate glass with white enamel marking
TRUSTED_SUSTAINABILITY
Specifications

Specifications

38 mm
Borosilicate Glass
5
Conical
105 mm
500 mL
183 mm
Culture Flask
Flat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers