missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ PVDF (Polyvinylidene Fluoride) Three Way Stopcocks

The three-way configuration of these stopcocks permits connection of any two or all three ports

Brand:  Bel-Art™ SP Scienceware™ F30888-0008

 View more versions of this product

Product Code. 15311681

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Barbed tubulations accept rubber or plastic tubing
  • Made of PVDF (polyvinylidene fluoride), a fluoroplastic resistant to most chemicals
  • Withstands temperatures up to 204°C
  • Maximum pressure rating 34.3 KPa (5 psi)
  • Steam autoclavable
Specifications

Specifications

6 mm
PVDF
34.3 KPa
204°C max.
1 Case
3-Way Stopcock
Autoclavable
8 mm
1
Rubber or Plastic Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.