Learn More
Welch Ilmvac™ Pump 2028 Vacuum Oil Free Fully Programmable
Brand: Welch Ilmvac™ 2028-03
This item is not returnable.
View return policy
Description
Self-cleaning 230V 50/60Hz 303mm (w) x 422mm (d) x 287mm (h) flow rate 29L/min, 3mbar Welch
Product Content Correction
Product Title
Welch Ilmvac™ Pump 2028 Vacuum Oil Free Fully Programmable >
Spot an opportunity for improvement?Share a Content Correction