missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ PU900 Series Medium Duty Polyurethane Gloves

Product Code. 15630934
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides good mechanical resistance to abrasion and cuts. Ansell PU900 Series Medium Duty Polyurethane Gloves provide comfort and dexterity.

  • Mechanical protection gloves
  • Medium duty cut resistance gloves
  • Good abrasion and cut resistance
  • Low lint glove
  • Good comfort and dexterity

Applications:

  • Assembly of white goods
  • Plastics injection and moulding
  • Assembling metal parts and components
  • Aeronautics
  • Finishing works
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003
Description Gauge: 13
Color White, Gray
Exterior Finish Palm Coated
Material Polyurethane
Product Type Mechanical Protection Gloves
Size 10
Style Construction: Knitted
Glove Cuff Style Knit
Liner Material Spandex Dyneema™
Packaging Type 12 pairs in a bag, 240 pairs in a carton
Series PU900
Length (Metric) 196 to 252 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.