missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Spatulas

Fisherbrand™ PTFE Spatulas are 5 mm thick.

Brand:  Fisherbrand™ 043.240

 View more versions of this product

Product Code. 15689850

  • 19.95 € / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
  • 5 mm Thick
Specifications

Specifications

1 Ea.
240 x 30 mm
240 mm
Fisherbrand™ PTFE Spatulas
PTFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers