missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Spatulas
Fisherbrand™ PTFE Spatulas are 5 mm thick.
Brand: Fisherbrand™ 043.120
This item is not returnable.
View return policy
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
- 5 mm Thick
Specifications
1 Ea. | |
120 x 12 mm | |
120 mm |
Fisherbrand™ PTFE Spatulas | |
PTFE |