missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PTFE Scoop

All parts coming into contact with the sample medium are made of pure white PTFE. Completely inert against all substances (with the exception of elementary fluoride). The rod is stabilised with a steel core which is completely encapsulated in PTFE.

Brand:  Buerkle™ 5389-0250

 View more versions of this product

Product Code. 10403522

  • 281.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Medium contacting parts: pure white PTFE
  • Completely inert against all substances (with the exception of elementary fluoride)
  • The rod is stabilised with a steel core which is completely encapsulated in PTFE
  • The 60 cm long rod can be extended to 120 cm by adding another component
  • The scoop PTFE with 10 ml has a strong rod with a length of 155 mm
Specifications

Specifications

Measuring Scoop
Autoclavable
Pure White
600 mm
PTFE
250 mL
95 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.