missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ PTFE and PPS Glass Stirrer Bearings

Suitable for vacuum use, these are perfect bearings for stirrer shafts of stainless steel and glass.

Brand:  Bohlender™ C425-06

Product Code. 15786425

  • 125.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Providing temperature resistance from −15° to +200°C
  • Universal chemical resistance
  • FDA conformity
  • Combination of a borosilicate glass piece with ground joint, an interior PTFE shaft guide with integrated special gasket and a GL screw cap of PPS
  • Special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap, thread 25
  • Provides a good sealing of the stirrer shaft
  • Gasket can be exchanged after wearing
Specifications

Specifications

Total length of 90 mm
EU NS 29/32
6 mm stirrer shafts
Glass Stirrer Bearings
GL 25 thread screw cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.