missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ PTFE Dipper

Ideal for obtaining surface samples from a wide range of liquids. Sampling Systems™ PTFE Dippers are quick and easy to use.

Brand:  Sampling Systems™ 5100A-1000

 View more versions of this product

Product Code. 15859361

  • 269.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PTFE is very durable and is resistant to chemical attack
  • PTFE product contact parts
  • Screw-in extension rods to increase the handle length
  • Suitable for use up to 280°C (400°F)
Specifications

Specifications

Dipper
Removable
Sampling Liquid
105 mm
PTFE
1000 mL
Up to 280°C
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.