Learn More
Testo™ Pt100 Laboratory Probe
Brand:  Testo™ 0609 7072
Product Code. 10029340
  • 241.00 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Probe, Laboratory
Standard 60751
45 sec.
-50°C to +400°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Testo™ Pt100 Laboratory Probe > Temperature Range: –50°C to 400°C