missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino Ni-TED 150 kit for His-tag Protein Purification in Mini format
The matrix of choice for highest protein purity in Mini format.
52.00 € - 200.00 €
Specifications
Bed Volume | 80 μL |
---|---|
Binding Capacity | 400μg |
Kit Contents | Protino Ni-TED 150 Packed Columns, buffers |
Matrix | Macroporous silica |
Content And Storage | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10316642
|
Macherey-Nagel™
745100.10 |
1 Kit |
52.00 €
Pack of 10 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
10707784
|
Macherey-Nagel™
745100.50 |
1 Kit |
200.00 €
Pack of 50 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
Description
Protino™ NI-TED, the purer alternative for His-tag protein purification
- Chelating group TED, one protein binding site
- High stability against reducing/chelating agents
- Low metal leaching
- High protein purity
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
Protino™ NI-TED packed columns
- Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
- Buffers included
Protino™ NI-TED resin
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-TED resin
- Empty 96 well receiver plates available for use with Protino™ Ni-TED resin
For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
80 μL | |
Protino Ni-TED 150 Packed Columns, buffers | |
Room Temperature | |
Purification of polyhistidine-tagged proteins | |
TED (tris-carboxymethyl ethylene diamine) |
400μg | |
Macroporous silica | |
1 Kit | |
Ni-TED Protein Purification Kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Protino Ni-TED 150 packed columns (10)
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360D-May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : imidazole, monosodium phosphate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title