missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Proteinase K

Delivered with a stabilizing solution for enhanced stability. Macherey-Nagel™ Proteinase K is lyophilized proteinase K supplied in pre-measured vials for easy preparation of 20mg/mL stock solution. It can be used with NucleoSpin™ and NucleoBond™ kits.

Brand:  Macherey-Nagel™ 740506.75


Product Code. 10655875

  • 85.90 € / 75mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Proteinase K is necessary for the isolation of DNA or simultaneous RNA / DNA isolation for DNA viruses (e.g. HBV) that are more difficult to lyse. Proteinase K treatment is also recommended for viral RNA isolation when viscous samples have to be processed (e.g., sputum samples).

  • Delivered with stabilizing solution to enable storage for up to six months at −20°C 
  • No decrease of proteinase activity when dissolved in the buffer compared to dissolving in water
  • Lyophilized mataerial is supplied in precisely measured vials, to which buffer solution is added to prepare 20mg/mL stock solution
  • Suitable for use with the NucleoSpin™ RNA Virus F kit and other NucleoSpin™ and NucleoBond™ kits, and also suitable for use in most other applications
Specifications

Specifications

20 mg/mL
Buffer solution
NucleoSpin™ and NucleoBond™ kits
Vial
Tritirachium album
Proteinase K
White
Viral RNA and DNA isolation, including: HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1
8mL Proteinase Buffer PB
Proteinase and Buffer, separately: room temperature (18–25°C) for at least 1 year
Prepared Stock Solution: −20°C for up to 6 months
75 mg
Lyophilized
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.