missing translation for 'onlineSavingsMsg'
Learn More

3M™ Hooded Anti-Static Protective Coveralls 4565 Series

Product Code. 14820992
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Comfortable coveralls made with high-performance fabric offer advanced quality and protection in challenging conditions such as exposure to hazardous dusts and particulates.

  • Type 4/5/6 Coverall designed to offer a very good barrier to hazardous particles and limited liquid splash and spray
  • Made of bicomponent nonwoven with ultra-low linting PE laminate for a noticeably comfortable drape
  • 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
  • Taped seams across the suit for increased hold out and strength reducing the risk of tearing and exposure
  • Protection against EN 14126 Biological agents
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out Til class 2
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Small
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Knit
Closure Type 2-way Zipper with Sealed Storm Flap
Seam Type Taped
Sterility Non-sterile
Coating Anti-static
Certifications/Compliance ASTM F1670, EN 14126:2003, EN 1149-1:2006, EN 13034, EN-ISO 13982-30
Description Elasticated Fitting, 3 Panel Hood, 2 Way Zipper, Knitted Cuffs, Low Linting Material
For Use With (Application) Pharmaceutical Processing, Chemical Manufacturing , Pesticide spraying
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'