missing translation for 'onlineSavingsMsg'
Learn More

Propiophenone, 99%, Thermo Scientific Chemicals

Product Code. 10715791
Click to view available options
Quantity:
2 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-55-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
InChI Key KRIOVPPHQSLHCZ-UHFFFAOYSA-N
Synonym propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
PubChem CID 7148
ChEBI CHEBI:425902
IUPAC Name 1-phenylpropan-1-one
SMILES CCC(=O)C1=CC=CC=C1

Specifications

Melting Point 18.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 218.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COC2H5
Refractive Index 1.5248 to 1.5268
Quantity 2 mL
Beilstein 07,II,231
Merck Index 15,7944
Specific Gravity 1
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with methanol, ether, benzene and toluene, insoluble in glycerol, ethylene glycol and, propylene glycol
Formula Weight 134.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propiophenone
Show More Show Less
Product Identifier
  • Propiophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'