missing translation for 'onlineSavingsMsg'
Learn More

Propiophenone, 99%, Thermo Scientific Chemicals

23.42 € - 204.02 €

Chemical Identifiers

CAS 93-55-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00009309
InChI Key KRIOVPPHQSLHCZ-UHFFFAOYSA-N
Synonym propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
PubChem CID 7148
ChEBI CHEBI:425902
IUPAC Name 1-phenylpropan-1-one
SMILES CCC(=O)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11422118
View Documents
Thermo Scientific Alfa Aesar
A15140.22
100 g
23.42 €
100g
Please to purchase this item. Need a web account? Register with us today!
11432118
View Documents
Thermo Scientific Alfa Aesar
A15140.36
500 g
49.45 €
500g
Please to purchase this item. Need a web account? Register with us today!
11442118
View Documents
Thermo Scientific Alfa Aesar
A15140.0E
2500 g
204.02 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Propiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

93-55-0
134.178
KRIOVPPHQSLHCZ-UHFFFAOYSA-N
7148
1-phenylpropan-1-one
C9H10O
MFCD00009309
propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
CHEBI:425902
CCC(=O)C1=CC=CC=C1

Specifications

93-55-0
1.008
87°C (188°F)
1.526
606215
propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
KRIOVPPHQSLHCZ-UHFFFAOYSA-N
1-phenylpropan-1-one
7148
134.18
Propiophenone
17°C to 18°C
217°C to 218°C
C9H10O
MFCD00009309
14,7830
Difficult to mix.
CCC(=O)C1=CC=CC=C1
134.178
CHEBI:425902
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P264b-P280-P305+P351+P338-P370+P378q-P501c

H227-H319

missing translation for 'einecsNumber' : 202-257-6

missing translation for 'rtecsNumber' : UG7175000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.