missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Profile Sensa XP™ Latex Gloves

Product Code. 12715785
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

High strength and excellent sensitivity. Nitritex™ Profile Sensa XP™ Latex Gloves are extra-length, powder-free examination gloves that measure 300mm/12 in., providing additional coverage past the wrist. Certified to Class I Medical and Cat 3 PPE standards.

  • Natural rubber latex provides exceptional strength while still providing excellent sensitivity
  • Mid-arm ambidextrous gloves measure 300mm for more coverage
  • Beaded cuff for easier donning and stability on the arm
  • Provides improved grip with a textured surface
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 455, EN 374; AQL 1,5
Classification Category III PPE, Class I Medical
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Hand Ambidextrous
Material Latex
Powdered Powder Free
Size Large
Sterility Non-sterile
Glove Cuff Style Beaded
Packaging Type Dispenser box
Quantity 100 Pk.
Length (English) Cuff 12 in.
Length (Metric) Cuff 300 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'