missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Profile PLHS Latex Gloves

Product Code. 12735805
Click to view available options
Size:
7
This item is not returnable. View return policy
This item is not returnable. View return policy

The Profile PLHS™ is the cost-effective sterile surgical glove. This 300mm/12 in. latex glove provides excellent flexibility and comfort, and is anatomically shaped to avoid wearer fatigue. Powder-free and featuring a beaded cuff for stability on the arm the PLHS also has a textured surface for excellent grip and grip

TRUSTED_SUSTAINABILITY

Specifications

Back Material Latex
Certifications/Compliance ISO 9001:2008 & 13485:2003 and Directive 93/42/EEC Annex V
Classification Class IIa Medical Device
Cuff Material Latex
Color Natural
Hand Specific
Material Latex
Powdered Powder Free
Product Type Gloves
Size 7
Sterility Sterile
Glove Cuff Style Beaded/Rolled
Packaging Type Boxed
Quantity 50 Pairs
Length (Metric) 305 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'