missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Profile™ PLHS Latex Gloves

Product Code. 12775805
Click to view available options
Size:
6
6.5
7.5
8
8.5
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

The cost-effective sterile surgical glove. Nitritex™ Profile™ PLHS Latex Glove is an extra-long latex glove with an anatomical shape that provides excellent flexibility and comfort to avoid wearer fatigue.

  • Made of powder-free Class IIa Medical grade latex
  • Measures 300mm/12 in. for more coverage than standard surgical gloves
  • Features a beaded cuff to ensure stability on the arm
  • Provides secure grip with a textured surface
TRUSTED_SUSTAINABILITY

Specifications

Back Material Latex
Certifications/Compliance ISO 9001:2008 & 13485:2003 and Directive 93/42/EEC Annex V
Classification Class IIa Medical Device
Cuff Material Latex
Color Natural
Hand Specific
Material Latex
Powdered Powder Free
Product Type Surgical Gloves
Size 9
Sterility Sterile
Glove Cuff Style Beaded/Rolled
Packaging Type Boxed
Quantity 50 Pairs
Length (Metric) 305 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'