Learn More
Contec ProChlor SATWIPES System
Point-of-use SATWIPE system, sterile ProChlor is a revolutionary sporicide achieving a 100% kill against spores in under 1 min.
Brand:  Contec XSWPP02PC
564.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 17809960
  • 795.00 € / Pack of 1200
Explore more special offers
Description
Description
  • Add 700ml of ProChlor to the dry roll, 5 mins before use, and use the presaturated wipes within 8 hours.
  • Meets requirements of Aseptic Transfer Guidelines
  • Log 5 kill against Bacillus subtilis in 60 seconds
  • Fast acting so suitable for transfer disinfection
  • Fast and easy to use
  • Very low residue, saving time on residue removal
  • Double bagged in linear tear packaging