Learn More
Zeiss Monitor integrated 5MP colour camera, live view tiltable TFT monitor 8.4'', transmitted light brightfield, phase contrast with objectives, condenser 0.4 Primo vert
Brand:  Zeiss 491206-0007-000
Monitor integrated 5MP colour camera, live view tiltable TFT monitor 8.4'', transmitted light brightfield, phase contrast with objectives, condenser 0.4 Primo vert
Code : 50
Additional Details : Weight : 0.01000kg
Product Code. 12302508