missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23827-1-AP-20UL
This item is not returnable.
View return policy
Description
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Specifications
PRDM10 | |
Polyclonal | |
Unconjugated | |
PRDM10 | |
KIAA1231, PFM7, PR domain containing 10, PRDM10, TRIS, Tristanin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
382066, 56980 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UTQ7, Q9NQV6 | |
Prdm10 | |
PRDM10 Fusion Protein Ag20605 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction