Learn More
PPP3CC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 55163-1-AP-150UL
 Ver más versiones de este producto
Código de producto. 16826645
  • 410.00 € / 150µL
Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones
Descripción
Descripción
This antibody is specific to PPP3CC.
Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca, calcineurin A-beta, and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).
Especificaciones
Especificaciones
PPP3CC
Polyclonal
Unconjugated
PPP3CC
36 53aa, CALNA3, CAM PRP catalytic subunit, CNA3, PPP3CC
Rabbit
Antigen Affinity Chromatography
RUO
19057, 5533
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P48454
Ppp3cc
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales