missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Powered Ice Sampler

Sampling Systems™ Powered Ice Sampler fits all standard electrical drills and takes samples in seconds.

Brand:  Sampling Systems™ 6010A-240

Product Code. 15819381

  • 479.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Chemically resistant 316L Stainless Steel construction
  • Fits all standard electric drills (drill not supplied)
  • Takes samples in seconds
  • Wide diameter for large cross sectional sampling
  • Supplied with ejection rod
Specifications

Specifications

Stainless Steel (316L)
Ice Sampling
240 mm
Discharge rod
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers