Learn More
Potassium trimethylsilanolate, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
82.91 € - 463.31 €
Chemical Identifiers
CAS | 10519-96-7, 109-99-9 |
---|---|
Molecular Formula | C3H10KOSi |
Molecular Weight (g/mol) | 129.30 |
MDL Number | MFCD00013421 |
InChI Key | COTHYYYVPUZALV-UHFFFAOYSA-N |
Synonym | potassium trimethylsilanolate, trimethylsiloxypotassium, silanol, trimethyl-, potassium salt, trimethylsilanol potassium salt, potassium trimethyl silanolate, silanol, 1,1,1-trimethyl-, potassium salt 1:1, kosime3, kotms, tmsok, c3h9osi.k |
PubChem CID | 23668039 |
SMILES | [K].C[Si](C)(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13458688
|
Thermo Scientific
448461000 |
100 mL | AcroSeal™ Glass bottle |
82.91 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13448688
|
Thermo Scientific
448468000 |
800 mL | AcroSeal™ Glass bottle |
463.31 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10519-96-7, 109-99-9 | |
129.30 | |
COTHYYYVPUZALV-UHFFFAOYSA-N | |
23668039 |
C3H10KOSi | |
MFCD00013421 | |
potassium trimethylsilanolate, trimethylsiloxypotassium, silanol, trimethyl-, potassium salt, trimethylsilanol potassium salt, potassium trimethyl silanolate, silanol, 1,1,1-trimethyl-, potassium salt 1:1, kosime3, kotms, tmsok, c3h9osi.k | |
[K].C[Si](C)(C)O |
Specifications
10519-96-7, 109-99-9 | |
(CH3)3SiOK | |
−17°C | |
COTHYYYVPUZALV-UHFFFAOYSA-N | |
potassium;trimethyl(oxido)silane | |
23668039 | |
0.91 | |
Potassium trimethylsilanolate |
C3H10KOSi | |
MFCD00013421 | |
potassium trimethylsilanolate, trimethylsiloxypotassium, silanol, trimethyl-, potassium salt, trimethylsilanol potassium salt, potassium trimethyl silanolate, silanol, 1,1,1-trimethyl-, potassium salt 1:1, kosime3, kotms, tmsok, c3h9osi.k | |
[K].C[Si](C)(C)O | |
129.30 | |
128.29 | |
0.9100g/mL |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May form explosive peroxides.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 234-062-7
- Potassium trimethylsilanolate, 2M sol in THF
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only