Learn More
Potassium thiosulfate hydrate, 96+%, extra pure, Thermo Scientific Chemicals
76.58 € - 283.41 €
Chemical Identifiers
CAS | 1264198-56-2 |
---|---|
Molecular Formula | K2O3S2 |
Molecular Weight (g/mol) | 190.31 |
MDL Number | MFCD00150405 |
InChI Key | FGRVOLIFQGXPCT-UHFFFAOYSA-L |
PubChem CID | 61501 |
SMILES | [K+].[K+].[O-]S([S-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10035213
|
Thermo Scientific Acros
388691000 |
100 g | Plastic bottle |
76.58 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10638044
|
Thermo Scientific Acros
388695000 |
500 g | Plastic bottle |
283.41 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
1264198-56-2 | |
190.31 | |
FGRVOLIFQGXPCT-UHFFFAOYSA-L | |
[K+].[K+].[O-]S([S-])(=O)=O |
K2O3S2 | |
MFCD00150405 | |
61501 |
Specifications
1264198-56-2 | |
6 to 9 (5% soln.) | |
96.0 to 104% (On dry substance) (Iodimetry) | |
K2S2O3·xH2O | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
[K+].[K+].[O-]S([S-])(=O)=O | |
190.31 | |
190.33 | |
Extra Pure | |
Potassium thiosulfate hydrate |
Colorless to Yellow | |
Liquid | |
K2O3S2 | |
MFCD00150405 | |
FGRVOLIFQGXPCT-UHFFFAOYSA-L | |
dipotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
61501 | |
≥96% | |
4% max. (150°C) |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Potassium thiosulfate hydrate
RUO – Research Use Only