missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium thiocyanate, Honeywell™
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Quantity | 2.5 kg |
Linear Formula | KSCN |
UN Number | NONH for all modes of transport |
Beilstein | 3594799 |
Formula Weight | 97.18g/mol |
Percent Purity | ≥98% (AT) |
Chemical Name or Material | Potassium thiocyanate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction