Learn More
Potassium tetrafluoroborate, 99%, extra pure, Thermo Scientific Chemicals
32.10 € - 105.00 €
Chemical Identifiers
CAS | 14075-53-7 |
---|---|
Molecular Formula | BF4K |
Molecular Weight (g/mol) | 125.90 |
MDL Number | MFCD00011395 |
InChI Key | AKEBROIVCDHVSD-UHFFFAOYSA-N |
Synonym | potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride |
PubChem CID | 518872 |
IUPAC Name | potassium tetrafluoroboranuide |
SMILES | [K+].F[B-](F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10780561
|
Thermo Scientific Acros
269220250 |
25 g | Plastic bottle |
32.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10503801
|
Thermo Scientific Acros
269220010 |
1 kg | Plastic bottle |
105.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
14075-53-7 | |
125.90 | |
AKEBROIVCDHVSD-UHFFFAOYSA-N | |
518872 | |
[K+].F[B-](F)(F)F |
BF4K | |
MFCD00011395 | |
potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride | |
potassium tetrafluoroboranuide |
Specifications
530.0°C | |
White to Gray | |
98.5% min | |
KBF4 | |
15,7804 | |
Solubility in water: 4.4g/L (20°C). Other solubilities: slightly soluble in boiling alcohol | |
[K+].F[B-](F)(F)F | |
125.90 | |
125.91 | |
Extra Pure | |
Potassium tetrafluoroborate, c.p. |
14075-53-7 | |
Micro-crystalline Powder | |
BF4K | |
MFCD00011395 | |
potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride | |
AKEBROIVCDHVSD-UHFFFAOYSA-N | |
potassium tetrafluoroboranuide | |
518872 | |
99% | |
2.5 |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 237-928-2
missing translation for 'rtecsNumber' : ED2800000
missing translation for 'tsca' : TSCA
RUO – Research Use Only