missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetraborate tetrahydrate, 99.5%, extra pure, Thermo Scientific Chemicals
35.88 € - 322.61 €
Chemical Identifiers
CAS | 12045-78-2 |
---|---|
Molecular Formula | B4K2O7·4H2O |
Molecular Weight (g/mol) | 305.5 |
Synonym | Potassium biborate tetrahydrate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11390766
|
Thermo Scientific Acros
259572500 |
250 g |
35.88 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10541301
|
Thermo Scientific Acros
259570010 |
1 kg |
97.28 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11964221
|
Thermo Scientific Acros
259570051 |
5 kg |
322.61 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12045-78-2 | |
B4K2O7·4H2O | |
305.5 | |
Potassium biborate tetrahydrate |
Specifications
850.0°C | |
White | |
99.5% | |
K2B4O7·4H2O | |
Potassium biborate tetrahydrate | |
305.5 | |
99.5% | |
Plastic bottle |
12045-78-2 | |
Powder | |
B4K2O7·4H2O | |
14,768 | |
Solubility in water: soluble. | |
305.5 | |
Extra Pure | |
Potassium tetraborate tetrahydrate |
Safety and Handling
GHS H Statement
Suspected of damaging the unborn child.
GHS P Statement
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium tetraborate tetrahydrate
Signal Word
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Potassium borate tetrahydrate
RUO – Research Use Only