missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfamate, 98%, Thermo Scientific Chemicals

43.82 € - 502.50 €

Chemical Identifiers

CAS 13823-50-2
Molecular Formula H2KNO3S
Molecular Weight (g/mol) 135.178
MDL Number MFCD00059941
InChI Key BTAAXEFROUUDIL-UHFFFAOYSA-M
Synonym potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
PubChem CID 23687442
IUPAC Name potassium;sulfamate
SMILES NS(=O)(=O)[O-].[K+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11485897
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24447.14
25 g
43.82 €
25g
Please to purchase this item. Need a web account? Register with us today!
11495897
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24447.22
100 g
131.66 €
100g
Please to purchase this item. Need a web account? Register with us today!
11405907
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24447.36
500 g
502.50 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

13823-50-2
135.178
BTAAXEFROUUDIL-UHFFFAOYSA-M
23687442
NS(=O)(=O)[O-].[K+]
H2KNO3S
MFCD00059941
potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
potassium;sulfamate

Specifications

13823-50-2
Crystalline Solid
H2NSO3K
potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
NS(=O)(=O)[O-].[K+]
135.178
135.18
Hygroscopic
White
H2KNO3S
MFCD00059941
BTAAXEFROUUDIL-UHFFFAOYSA-M
potassium;sulfamate
23687442
98%
Potassium sulfamate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only