Learn More
Potassium pyrosulfate, 98%, for analysis, Thermo Scientific Chemicals
80.20 € - 293.00 €
Chemical Identifiers
CAS | 7790-62-7 |
---|---|
Molecular Formula | K2O7S2 |
Molecular Weight (g/mol) | 254.31 |
InChI Key | KAQHZJVQFBJKCK-UHFFFAOYSA-L |
Synonym | potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat |
PubChem CID | 62681 |
IUPAC Name | dipotassium;sulfonato sulfate |
SMILES | [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10022640
|
Thermo Scientific Acros
207915000 |
500 g | Plastic bottle |
80.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914251
|
Thermo Scientific Acros
207910025 |
2.5 kg | Plastic bottle |
293.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
7790-62-7 | |
254.31 | |
potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat | |
dipotassium;sulfonato sulfate |
K2O7S2 | |
KAQHZJVQFBJKCK-UHFFFAOYSA-L | |
62681 | |
[O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+] |
Specifications
325.0°C | |
White | |
98% | |
K2S2O7 | |
potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat | |
KAQHZJVQFBJKCK-UHFFFAOYSA-L | |
dipotassium;sulfonato sulfate | |
62681 | |
98% | |
2.51 | |
Potassium pyrosulfate |
7790-62-7 | |
Fine Crystalline Powder | |
K2O7S2 | |
15,7784 | |
Solubility in water: may decompose | |
[O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+] | |
254.31 | |
254.31 | |
Analytical | |
2.5100g/mL |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Corrosive to the respiratory tract.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-216-8
- Potassium pyrosulfate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only